Ward's® Diffusion and Cell Size Lab Activity
Catalog #: 470178-380
Supplier: Avantor
Specifications
- Description:Ward's® Diffusion and Cell Size Lab Activity
- Cat. no.:470178-380
- Supplier No.:WARD470178-380
Specifications
About this item
Visualize an abstract concept in under fifteen minutes.
- No titrations needed
- Use safe solutions throughout the activity
- Hands-on learning activity
- Pre-made agar cubes included
Students simulate 'cells' using agar blocks to illustrate the relationship between the size of the cell and its volume. In the process, students predict outcomes by varying the 'cell' size, measure the rates of diffusion, and learn about factors that affect the rates of diffusion across a cell membrane. This innovative procedure requires no titrations and uses completely safe solutions. The demonstration size contains enough materials for five demonstrations.
Ordering information: At times, supply may be limited due to high demand. Please place your order approximately two weeks in advance. Note: Coupon included for perishable materials. Redeem by mail, fax, phone, or e-mail.
Customers Who Bought This Also Bought