Iron-Deficiency Anemia Slide
Catalog #: 470177-660
Supplier: Avantor
Specifications
- Description:Iron-Deficiency Anemia Slide
- Cat. no.:470177-660
- Stain:Giemsa
- Supplier No.:WARD470177-660
- Orientation:Smear
Specifications
About this item
Illustrates red blood cell characteristics of the deficiency
- Identifying features clearly distinguishable
- Rigorous quality control standards
Human. Peripheral blood. Characterized by small, pale red blood cells.
Customers Who Bought This Also Bought