Human Blood Slide
Catalog #: 470177-632
Supplier: Avantor
Specifications
- Description:Human Blood, smear
- Cat. no.:470177-632
- Stain:Wright's
- Supplier No.:WARD470177-632
- Orientation:Smear
Specifications
About this item
Smear, Wright's stain
- All identifying features are clearly distinguishable
- Rigorous quality control standards
Shows the three major functional classes: red cells, white cells, and platelets.
Customers Who Bought This Also Bought