Cardiac Muscle, Intercalated Discs Slide
Catalog #: 470177-594
Supplier: Avantor
Specifications
- Description:Cardiac Muscle, Intercalated Discs
- Cat. no.:470177-594
- Stain:Iron Hematoxylin
- Supplier No.:WARD470177-594
- Orientation:Section
Specifications
About this item
Illustrates cardiac muscle characteristics
- Identifying features clearly distinguishable
- Rigorous quality control standards
Mammal. Stained to demonstrate intercalated discs and approximate central location of nuclei in fibers.
Customers Who Bought This Also Bought