Brine Shrimp Eggs
Catalog #: 470152-880
Supplier: Avantor
Specifications
- Description:Brine Shrimp Eggs
- Cat. no.:470152-880
- Supplier No.:WARD470152-880
Specifications
About this item
Place these eggs in a brine solution for two days in order to make the shrimp (Artemia) hatch. Our special processing technique allows you to store the eggs for nearly indefinite periods of time. 7 grams.
Customers Who Bought This Also Bought