Order Entry
ContactUsLinkComponent
CYLINDER SQUAT 50 ML GRADUATED PLASTIC
Wide-Mouth Polypropylene Cylinders
Catalog #: 470150-424
Supplier:  SI Manufacturing
Wide-Mouth Polypropylene Cylinders
Catalog #: 470150-424
Supplier:  SI Manufacturing

Specifications

  • Capacity:
    50 ml
  • Cat. no.:
    470150-424
  • Supplier No.:
    78110

Specifications

About this item

Chemical and Heat Resistant to 80°C.

  • Wide mouth is ideal for performing density tests
  • Molded, blue over-printed graduations
  • Aligned with GLOBE protocols

The squat design increases stability on the bench while the low form requires less overhead storage space.

Customers Who Bought This Also Bought