Order Entry
ContactUsLinkComponent
RECYCLING RIKER MOUNT
Recycling Riker Mount
Catalog #: 470028-108
RECYCLING RIKER MOUNT
Recycling Riker Mount
Catalog #: 470028-108

Specifications

  • Length:
    12"
  • Width:
    16"
  • Description:
    Recycling Riker Mount
  • Cat. no.:
    470028-108
  • Supplier No.:
    E900
  • No. of Parts:
    1

Specifications

About this item

Discover Better Uses for Common Waste Materials

This unique mount displays materials that we use in our everyday lives with text descriptions highlighting the common sources and what they can be recycled into. Materials included in the display are plastics, wood, textiles, rubber, batteries, metals, glass, paper, and aggregates. Size: 12" x 16".

Customers Who Bought This Also Bought