Scoop Type Spatula
Catalog #: 470177-948
Supplier: United Scientific Supplies
Specifications
- Description:Laboratory scoop (blade only)
- Supplier No.:LSCSS6
Specifications
About this item
This scoop-type spatula is versatile and durable.
- Spatula dimensions: ¹/₂×7¹/₂"
- One rounded end, one pointed end
The stainless steel spatula has a semicircular cross section.
Customers Who Bought This Also Bought