Mammalian Ovary
Catalog #: 470177-012
Supplier: Avantor
Specifications
- Description:Ovary (Mammalian), Mature Follicles
- Cat. no.:470177-012
- Stain:Mallory triple
- Supplier No.:WARD470177-012
- Orientation:Cross Section
Specifications
About this item
Observe the graafian follicle in different stages.
- Identifying features clearly distinguishable
- Rigorous quality control standards
Mammal. Cat ovary showing graafian follicles. Selected to show germ hill and large ovum.
Ovary (Mammalian), cross section- Demonstrates developing follicles
Ovary (Mammalian), cross section- Shows maturing follicle
Customers Who Bought This Also Bought