Marchantia Life History, Composite Slide
Catalog #: 470176-860
Supplier: Avantor
Specifications
- Description:Marchantia Life History, Composite Slide
- Cat. no.:470176-860
- Stain:Quadruple stained
- Supplier No.:WARD470176-860
- Orientation:Section
Specifications
About this item
Liverwort life stages comparison
- Identifying features clearly distinguishable
- Rigorous quality control standards
Liverwort. Shows gemmae cup, antheridia, archegonia, and sporophyte.
Customers Who Bought This Also Bought