ContactUsLinkComponent
PANDORINA MORUM 100 STUDENTS
Pandorina
Catalog #: 470176-700
Supplier:  Avantor
PANDORINA MORUM 100 STUDENTS
Pandorina
Catalog #: 470176-700
Supplier:  Avantor
Specifications
  • Description:
    Pandorina Culture for 100 Students
  • Cat. no.:
    470176-700
  • Supplier No.:
    WARD470176-700
Specifications
About this item
Freshwater. Colonial; colonies elliptical, made up of 16–32 cells. Basic culture solution.
Characterized by green plastids and cell walls composed of cellulose.
Customers Who Bought This Also Bought