Pandorina
Catalog #: 470176-698
Supplier: Ward's Science
Specifications
- Description:Pandorina morum (Small)
- No. of Students:30
- Cat. no.:470176-698
- Packaging:Tube
- Media:Basic culture solution
- Supplier No.:WARD470176-698
Specifications
About this item
Freshwater. Colonial; colonies elliptical, made up of 16–32 cells. Basic culture solution.
Characterized by green plastids and cell walls composed of cellulose.
Customers Who Bought This Also Bought