Ward's® Live Gloeocapsa Culture
Catalog #: 470176-682
Supplier: Ward's Science
Specifications
- Description:Gloeocapsa (Small)
- No. of Students:30
- Cat. no.:470176-682
- Packaging:Tube
- Media:Basic culture solution
- Supplier No.:WARD470176-682
Specifications
About this item
This colonial cyanobacteria is structured as several cells bound together in a gelatinous "mucilaginous sheath".
Commonly found in masses on wet rocks in aquatic or semi-aquatic habitats.
Genus: Gloeocapsa Species: Not specified
Customers Who Bought This Also Bought