Scenedesmus
Catalog #: 470176-650
Supplier: Ward's Science
Specifications
- Description:Scenedesmus (Small)
- No. of Students:30
- Cat. no.:470176-650
- Packaging:Tube
- Media:Basic culture solution
- Supplier No.:WARD470176-650
Specifications
About this item
Freshwater. Colonial; four cells with spines. Basic culture solution.
Characterized by green plastids and cell walls composed of cellulose.
Customers Who Bought This Also Bought