Live Physarum polycephalum Culture
Catalog #: 470176-642
Supplier: Ward's Science
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Description:Physarum polycephalum - Plasmodial Stage
- No. of Students:30
- Cat. no.:470176-642
- Packaging:Plate
- Media:Physarum-oat agar
- Supplier No.:WARD470176-642
Specifications
About this item
This "many-headed" organism is a plasmodial slime mold commonly found in forests on decaying deciduous trees. Available in both plasmodial and sclerotial stages.
- Biosafety Level: 1
- The Plasmodial Stage is the active feeding stage of Physarum and consists of a network of protoplasmic veins.
- Plasmodial Stage is excellent for viewing Protoplasmic Streaming, or "Shuttle streaming"
- The Sclerotial Stage is comprised of macrocysts, and is Physarum's dormant stage. It protects the organism until it is able to rehydrate in the presence of a food source.
Ordering information: Bacterial and fungal cultures are restricted and can only be ordered by educational and research professionals. Ward's Science is not affiliated with the American Type Culture Collection and does not provide corresponding ATCC numbers for our organisms. All fungal cultures undergo an extensive quality control program prior to releasing for sale, so they are guaranteed for purity as well as for species. This organism is offered as a ready-to-use plate culture, as well as a dormant, dry version that can be rehydrated for culture at a later time.
Delivery: These products contain living or perishable materials and will ship via overnight or 2nd day delivery. Please specify a delivery date during checkout.
Caution: Though this organism is not considered to be pathogenic, Ward's recommends that you utilize proper aseptic technique when handling all microorganisms.
Customers Who Bought This Also Bought