Ward's® Live Freshwater Rotifer Culture
Catalog #: 470176-594
Supplier: Avantor
Specifications
- Description:Freshwater Rotifers (Large culture)
- Storage Conditions:Room temp, loose cap
- No. of Students:100
- Cat. no.:470176-594
- Packaging:3 Tubes
- Media:Hay culture media
- Supplier No.:WARD470176-594
Specifications
About this item
Study the Rotifera under your microscope to discover the variety of structures among them. Subculture them in Alfalfa or Hay medium to increase your supply. Species may vary based on availability.
Rotifers have a wide variety of movement types ranging from free moving to sessile. They can be found in any healthy freshwater lakes and ponds.
Customers Who Bought This Also Bought