To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Description:Kocuria rhizophila (Micrococcus luteus)
- Cat. no.:470176-534
- Packaging:Tube
- Media:Tryptic soy agar
- Supplier No.:WARD470176-534
- Incubation Conditions:25 °C
Specifications
About this item
Kocuria rhizophila is also commonly known by the name Micrococcus luteus. This organism is commonly isolated from soil, and is frequently used to test antimicrobial efficacy in the food industry.
- Biosafety Level: 1
- Gram Stain: Positive
- Morphology: Cocci in tetrads
- Motility: Negative
- Respiration: Aerobic
- Chromogenesis: Yes (yellow)
All bacterial cultures undergo an extensive quality control program prior to releasing for sale, so they are guaranteed for purity as well as for species.
This organism is offered as a ready-to-use tube culture, as well as in a freeze-dried culture for longer storage.
Freeze-dried organisms come with everything you need to rehydrate and culture the organism when you're ready.