To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Description:B. subtilis: Freeze dried with media pack
- Storage Conditions:Freezer
- Cat. no.:470176-526
- Packaging:Lyophilized
- Media:Tryptic soy agar
- Supplier No.:WARD470176-526
Specifications
About this item
This organism is commonly isolated in soil and produces antibiotics such as bacitracin, subtilisin, and mycobacillin.
- Biosafety Level: 1
- Gram Stain: Positive
- Morphology: rods in chains
- Motility: Positive
- Respiration: Aerobic
- Spores: Yes, centrally located
All bacterial cultures undergo an extensive quality control program prior to releasing for sale, so they are guaranteed for purity as well as for species.
It's gram stain is positive, and its spores are centrally located.