Cladophora
Catalog #: 470176-496
Supplier: Ward's Science
Specifications
- Description:Cladophora (Small)
- No. of Students:30
- Cat. no.:470176-496
- Packaging:Tube
- Media:Erdschreiber's medium
- Supplier No.:WARD470176-496
Specifications
About this item
Marine. Filamentous; freely branched. Erdschreiber’s medium.
Characterized by green plastids and cell walls composed of cellulose.
Customers Who Bought This Also Bought