Live Amoeba proteus cultures
Catalog #: 470176-446
Supplier: Avantor
Specifications
- Description:Amoeba proteus Vital Stained Culture (small)
- Storage Conditions:Room temp, loose cap
- No. of Students:30
- Cat. no.:470176-446
- Media:Amoeba medium
- Supplier No.:WARD470176-446
Specifications
About this item
Demonstrate amoeba structure, locomotion, and feeding behavior.
- Demonstrate amoeba structure, locomotion, and feeding behavior
- ReadySlides are prepared for you you and offer a convenient, ready-to-go option
- Vital Stained cultures allow students to better visualize organism structures
Amoeba proteus is a great sarcodine for demonstrating amoebae structure. They use pseudopods for both locamotion and feeding. Amoeba feed on Chilomonas, or other small protozoa, which they phagocytize with their pseudopodia. Amoeba cultures are supplied in Amoeba Media with a rice grain and food (Chilomonas).
Ordering information: These products contain living or perishable materials. Please specify a delivery date.
Customers Who Bought This Also Bought