Ward's® Peridinium
Catalog #: 470176-434
Supplier: Avantor
Specifications
- Description:Peridinium Culture for 100 Students
- Cat. no.:470176-434
- Supplier No.:WARD470176-434
Specifications
About this item
Freshwater. Unicellular. Soil-water medium. Characterized by two flagella (one inside a groove at right angles to the other); most also have tests made of cellulose and are encrusted with silica.
Customers Who Bought This Also Bought