Spirulina
Catalog #: 470176-414
Supplier: Avantor
CAS Number:
Specifications
- Description:Spirulina Marine Medium
- Size:120 Students
- Cat. no.:470176-414
- Supplier No.:WARD470176-414
Specifications
About this item
Marine. Unicellular. Food organism, brackish, helically coiled filaments. Marine medium.
Shipped in a form that provides the best quality specimen. Most are supplied in 16 x 125 mm tubes, 2 oz. jars, or 8 oz. jars.
Customers Who Bought This Also Bought