Spirulina
Catalog #: 470176-412
Supplier: Ward's Science
CAS Number:
Specifications
- Description:Spirulina Marine Medium
- Size:30 students
- Cat. no.:470176-412
- Supplier No.:WARD470176-412
Specifications
About this item
Marine. Unicellular. Food organism, brackish, helically coiled filaments. Marine medium.
Shipped in a form that provides the best quality specimen. Most are supplied in 16 x 125 mm tubes, 2 oz. jars, or 8 oz. jars.
Customers Who Bought This Also Bought