Order Entry
ContactUsLinkComponent
LIVE COLIPHAGE T4 VIRAL CULTURE
Ward's® Live Coliphage Suspensions
Catalog #: 470176-272
Supplier:  Avantor
Ward's® Live Coliphage Suspensions
Catalog #: 470176-272
Supplier:  Avantor
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Description:
    Coliphage: T4
  • Storage Conditions:
    Refrigerator
  • Cat. no.:
    470176-272
  • Packaging:
    Vial
  • Media:
    Tryptic soy broth
  • Supplier No.:
    WARD470176-272
  • Incubation Conditions:
    37 °C

Specifications

About this item

A collection of some of the more popular forms of coliphage. The successful culture of coliphage requires growing in the appropriate host bacterium (available separately)

Liquid coliphage suspension are routinely tested for quality, purity, and titer. They contain DNA or RNA supplied as a filtered tryptic soy broth suspension of relative infective titer (10-7 plaque-forming units per mL). Several varieties of coliphage cultures are available.

Ordering information: Contents include: PhiX174: Minute polyhedral form, 25 ?m in dia. Circular single-stranded DNA close relative of S13 phage. Good for plaque morphology and demonstration of cations on plaque growth. T2r+: Wild type. Used most frequently in genetic studies. Rapid lysis. T4r+: Wild type. Less sensitive to ultraviolet radiation than T2. Rapid lysis. T4: Non-reverting plaque morphology. Mutant of T4r+. Rapid lysis. T4rIIA: Point mutation in cistron A, segment 4. Reversion rate to wild type approximately 106. T7: Rapid lysis. Produces large plaques. The liquid suspension is in tryptic soy broth. It is supplied in a vial and the proven titer is noted on the label. Quantity: 1.5 ml.
Customers Who Bought This Also Bought