Ward's® Live Mealworm Larvae, Pupae and Beetles (Tenebrio)
Catalog #: 470176-238
Supplier: Avantor
Specifications
- Description:Mixed Staged Set (50 total)
- Cat. no.:470176-238
- Supplier No.:WARD470176-238
Specifications
About this item
Teach your students about the complete metamorphosis of an insect with these mealworms.
- Store mealworms in bran until ready to use
- Excellent food source for turtles, salamanders and fish
- Demonstrates complete metamorphosis
Ordering information: Culture Kit includes culture pan, tenebrio nutrient, and a set of mixed stages as a starter culture.
Delivery: These products contain living materials and will ship via overnight or 2nd day delivery. Please specify a delivery date during checkout.
Packaging: Larvae and beetle cultures come packed with some tenebrio nutrient.
Customers Who Bought This Also Bought