Lil’pet Micropipette
Catalog #: 470149-114
Supplier: VWR International
Specifications
- Volume:10 µl
- Cat. no.:470149-114
- Supplier No.:MM-10
Specifications
About this item
Lil’pet Micropipettes feature ideal volumes for gel loading.
- Units accept standard micropipette tips
- Economical
- Durable
- Autoclavable
These fixed-volume pipettes feature a double stop plunger to expel all liquid from the tip.
The 1000 μl model includes two tips. All other pipettes come with one each.
Customers Who Bought This Also Bought