Porcelain Pestles
Catalog #: 470149-016
Supplier: SILICO & CHEMICO PORCELAIN SE
Specifications
- Description:Porcelain Pestle, 130 mm, for 80 mm Mortar (62615-27) 60314
- Cat. no.:470149-016
- Supplier No.:12091-260
Specifications
About this item
Pestles for Porcelain Mortars.
The end of each pestle is unglazed.
Customers Who Bought This Also Bought