To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Garment application:Chemical protection
- Gender:Unisex
- Color:Black
- L×W:42×27"
- Fabric:Extra-heavy rubber and cotton tabs
- Cat. no.:470148-648
- Type:Rubber apron
- Supplier No.:RBAP-AD
Specifications
About this item
Durable, lightweight, rubber apron resistant to chemicals, acids, and caustics.
- Chemical-resistant
- Extra-heavy rubber material
- Lightweight
- Contains latex
Extra-heavy rubber coated sheeting plus cotton tabs on the waist and neck have made these aprons a durable lab standard for years. They resist most chemicals, acids, and caustics, yet are still lightweight.