Economy Short Stem Funnels, Hutzler
Catalog #: 470150-352
Supplier: Hutzler Manufacturing
Specifications
- Description:Economy Short Stem Funnels, Top ID, mm: 70, Stem Length, mm: 19
- Cat. no.:470150-352
- Supplier No.:802
Specifications
About this item
Funnels are made of durable plastic. Funnels of top inner diameter 70-108 are polypropylene and autoclavable. Funnels with top inner diameter of 127 and 165 are polyethylene and not autoclavable.
The 165 mm size has a wide stem for quick transfers.
Customers Who Bought This Also Bought