Order Entry
ContactUsLinkComponent
RUBBER BULB - 30ML
Heavy Rubber Pipette Bulbs
Catalog #: 470149-454
Supplier:  VWR International
RUBBER BULB - 30ML
Heavy Rubber Pipette Bulbs
Catalog #: 470149-454
Supplier:  VWR International

Specifications

  • Capacity:
    30 ml
  • Description:
    Heavy Rubber Pipette Bulbs
  • Cat. no.:
    470149-454
  • Bore Size:
    5 mm
  • Supplier No.:
    CP1062/6

Specifications

About this item

Customers Who Bought This Also Bought