Order Entry
ContactUsLinkComponent
SPATULA S/S DBL/TAPERED BLADE
Nickel/Stainless Steel Spatulas, Double Bladed
Catalog #: 470149-448
Supplier:  VWR International
SPATULA S/S DBL/TAPERED BLADE
Nickel/Stainless Steel Spatulas, Double Bladed
Catalog #: 470149-448
Supplier:  VWR International

Specifications

  • Description:
    Nickel/stainless steel spatulas, double bladed, blade ends: one rounded, one tapered
  • Cat. no.:
    470149-448
  • Supplier No.:
    CH12222A/1

Specifications

About this item

Spatulas with 50×7 mm thin flat rounded blades on either end.

Wire Ø: 3 mm
Length: 8"

Customers Who Bought This Also Bought