Cuvette Rack
Catalog #: 470149-208
Supplier: VWR International
Specifications
- Description:Cuvette Rack
- Cat. no.:470149-208
- Supplier No.:260010
Specifications
About this item
Accommodate Most Square Cuvettes.
- Reduced risk of spilling
- Avoids risk of confusion with individually numbered spots
- Firmly holds up to 20 individual cuvettes
Customers Who Bought This Also Bought