CRUCIBLE LOW FORM SIZE 0 17 ML PORC
Catalog #: 470149-072
Supplier: SILICO & CHEMICO PORCELAIN SE
Customers Who Bought This Also Bought
Ethylene glycol ≥99%, Laboratory Reagent, VWR Chemicals BDH®
Supplier: VWR International
$321.66 to $979.57
Pentane ≥98.0%, Laboratory Reagent, VWR Chemicals BDH®
Supplier: VWR International
$500.19 to $1,790.36
Muscovite Mica Sheets, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
$69.52 to $88.34
Sodium chloride 99.0-100.5% (dried basis), granular USP, Multi-Compendial, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
$1,055.16
SP Bel-Art Fritware® Porous Polyethylene Sheets, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
$401.76
Clarity Urocheck Urinalysis Strips, Clarity Diagnostics
Supplier: Clarity Diagnostics
$31.63 to $756.61