Order Entry
ContactUsLinkComponent
VWR VIAL W/CAP CAPSULE 24/PK
Glass Vials with Screw Caps
Catalog #: 470146-668
Supplier:  Glass Vials
VWR VIAL W/CAP CAPSULE 24/PK
Glass Vials with Screw Caps
Catalog #: 470146-668
Supplier:  Glass Vials

Specifications

  • Cap included:
    Yes
  • Capacity:
    20.00 ml
  • Cat. no.:
    470146-668
  • Diameter ×H:
    27×57 mm
  • Supplier No.:
    V2757C-FM-SP

Specifications

About this item

Ideal for a variety of uses.

  • Accompanied by a polypropylene foam lined closed top cap
  • Cap designed with a continuous thread 'CT' finish
  • Creates a positive seal
  • Available in a wide range of sizes

Display specimens or keep your science supplies organized with these airtight glass vials featuring polypropylene caps.

Customers Who Bought This Also Bought