ContactUsLinkComponent
COMPRESSED AIR DISPENSER 12OZ EQUIP
Compressed Air Dispenser
Catalog #: 470146-262
Supplier:  STONER
COMPRESSED AIR DISPENSER 12OZ EQUIP
Compressed Air Dispenser
Catalog #: 470146-262
Supplier:  STONER
Specifications
  • Description:
    Compressed air dispenser, 12 oz.
  • Cat. no.:
    470146-262
  • Supplier No.:
    94203
Specifications
About this item
Ozone safe compressed air dispenser
Concentrated, directed jets of moisture-free filtered air safely blow dust particles from delicate instruments.
Customers Who Bought This Also Bought