Pipette Support
Catalog #: 470145-334
Supplier: Bel-Art
Specifications
- Description:Pipette Support
- Cat. no.:470145-334
- Supplier No.:F18953-0000
Specifications
About this item
Sturdy molded polypropylene rack holds pipettes up to 1/2" in dia in 5 x 10 arrangement.
Lower deck keeps pipette tips from surface beneath. Convenient hand grips on both sides. Autoclavable at 121 °C.
Size: 8-3/4" H x 8-1/2" W x 4-1/2" D".
Customers Who Bought This Also Bought