Order Entry
ContactUsLinkComponent
RAINBOW TAPE-.5 X500 PK 10 ASST. COLORS
Rainbow Laboratory Tape
Catalog #: 470144-262
Supplier:  NEVS INK INC SE
RAINBOW TAPE-.5 X500 PK 10 ASST. COLORS
Rainbow Laboratory Tape
Catalog #: 470144-262
Supplier:  NEVS INK INC SE

Specifications

  • Description:
    Rainbow Laboratory Tape, 1/2" x 500"
  • Cat. no.:
    470144-262
  • Supplier No.:
    VWREDU-T14

Specifications

About this item

Strong, durable, oil resistant, waterproof, and acid resistant, this label tape is fully equipped for laboratory use.

  • 1" core

It can withstand temperatures from -23 to +121°Cand -10 to +250°F. Easily write on the tape with pen, pencil, or permanent marker. It will stick to most surfaces including glass, plastic, and metal; yet it removes cleanly and easily. Includes 10 rolls. Size of each roll: 1/2" x 500".

Customers Who Bought This Also Bought