Convex Flask Brushes
Catalog #: 470123-992
Supplier: Mill-Rose
Specifications
- Bristle Color:White
- Bristle Material:Hog hair
- Handle Material:Plastic
- Description:Convex Flask Brushes
- Total length:356 mm
- Cat. no.:470123-992
- Supplier No.:2131
Specifications
About this item
White hog bristles mounted on a durable, plastic handle. Flexible brush section adapts to curvature of flasks.
Customers Who Bought This Also Bought