Order Entry
ContactUsLinkComponent
BOTTLE WASH W/SPRAYER - 16OZ
Washing Bottle with Trigger Sprayer
Catalog #: 470123-944
Supplier:  Tolco
Washing Bottle with Trigger Sprayer
Catalog #: 470123-944
Supplier:  Tolco

Specifications

  • Capacity:
    480 ml (16 oz.)
  • Description:
    Washing Bottle with Trigger Sprayer
  • Cat. no.:
    470123-944
  • Supplier No.:
    120112110412

Specifications

About this item

Great for general lab use.

  • Round polyethylene bottle with threaded neck
  • Polypropylene screw-on, trigger-style sprayer
  • Knurled nozzle adjustable from fine mist to jet stream
Customers Who Bought This Also Bought