ContactUsLinkComponent
SUPPORT T-NESTING FOR 13MM ROD
T-Nesting Support Bases
Catalog #: 470123-486
Supplier:  VWR International
T-Nesting Support Bases
Catalog #: 470123-486
Supplier:  VWR International
Specifications
  • Description:
    T-Nesting Support Bases, T Size L x W, in.: 7-1/8 x 8-3/4, Thread Size: 3/8 - 16, Rod Dia., in.: 1/2
  • Cat. no.:
    470123-486
  • Supplier No.:
    WLS78355-10C
Specifications
About this item
Specially designed to provide excellent stability, these bases are shaped with legs that slide freely under one another for compact storage or set up.
Customers Who Bought This Also Bought