Tripod Bases
Catalog #: 470123-480
Supplier: VWR International
Specifications
- Description:Tripod Base, 4" Radius, 3/8–16" Thread
- Cat. no.:470123-480
- Supplier No.:WLS78335-BY
Specifications
About this item
These heavy cast iron bases provide a three-point support for stability.
Customers Who Bought This Also Bought