ContactUsLinkComponent
STOPWATCH MECHANICAL - ECONOMY
Analog Stopwatch
Catalog #: 470123-440
Supplier:  VWR International
STOPWATCH MECHANICAL - ECONOMY
Analog Stopwatch
Catalog #: 470123-440
Supplier:  VWR International
Specifications
  • Description:
    Analog stopwatch
  • Cat. no.:
    470123-440
  • Supplier No.:
    PH0358A
Specifications
About this item
Count up to 29:59 with resolution of ¹/₅ sec. It is nickel-plated and features two-dials and a hard carrying case.
Size: 2"Ø.
Customers Who Bought This Also Bought