Variable Volume Economy Micropipettes
Catalog #: 470123-186
Supplier: United Scientific Supplies
Specifications
- Volume:100 - 1000 µl
- Description:1000 μL Micropipette
- Increments:1.0 µl
- Supplier No.:PVV-1000
Specifications
About this item
High performance pipette.
- Compatible with Gilson and eppendorf brand tips
- Built-in tip ejector
- Click stop digital system
- Easy calibration and maintenance
- Autoclavable tipcone assembly
Easy to disassemble for servicing and autoclaving. The 100 to 1000 μl micropipette uses 1000 μl tips. All other micropipettes use standard 200 μl tips.
Customers Who Bought This Also Bought