Dehydrated Media
Catalog #: 470113-152
Supplier: Culture Media & Supplies
CAS Number:
Specifications
- Packaging:Bag
- Description:Dehydrated Media Yeast Extract (component)
- Size:114 g
- Cat. no.:470113-152
- Supplier No.:LD184A
Specifications
About this item
Dehydrated Media is suitable for almost all educational applications.
- For Use with Plates, Slants, and Broth Media
- Instructions Included
- Sealed Bottle
- Expiration Date on Bottle
It arrives in a sealed bottle with an expiration date and complete instructions for preparation. Once items have shipped, they have three or more years before expiration.
Agar and components are used for plates or slants. Assuming a 20 mL fill for a 100 x 15 mm petri dish, each 114 g bottle will fill at least 100 dishes; each 454 g bottle will fill at least 400. Broth media are used for tube cultures. Assuming a 12 mL fill for a 20 x 75 mm tube, each 114 g bottle will fill at least 1000 tubes; each 454 g bottle will fill at least 2000.
Customers Who Bought This Also Bought