ContactUsLinkComponent
LID LOCK GUARDS MULTI PK100
Microcentrifuge Tube Lid Locks
Catalog #: 470112-976
LID LOCK GUARDS MULTI PK100
Microcentrifuge Tube Lid Locks
Catalog #: 470112-976
Specifications
  • Description:
    Microcentrifuge Tube Lid Locks, Mulit color with handle
  • Cat. no.:
    470112-976
  • Supplier No.:
    145063
Specifications
About this item
Secure caps to lids of microcentrifuge tubes with these autoclavable locks.
  • Easy to use tabs
  • Autoclavable
Each features a tab, allows to pick them out of a waterbath without burning your fingers.
Customers Who Bought This Also Bought