Order Entry
ContactUsLinkComponent
MICROSCOPE SLIDE 1CAVITY PK12
Student Grade Economy Microscope Slides
Catalog #: 470112-502
Supplier:  VWR International
Student Grade Economy Microscope Slides
Catalog #: 470112-502
Supplier:  VWR International

Specifications

  • Description:
    Single cavity slides, soda lime glas
  • L×W:
    3×1"
  • Thickness:
    1.0 - 1.2 mm
  • Cat. no.:
    470112-502
  • Supplier No.:
    BI0086A

Specifications

About this item

These economical slides are available in a variety of types.

  • Various Slide Types for All Needs
  • Standard Slide Size
  • Optically Clear Glass Construction
  • Ground-Glass Edges

They’re perfect for consistently reliable mounts without the high cost of premium grade slides.

The slides are made of optically clear glass with ground-glass edges.

Customers Who Bought This Also Bought