Edvotek® Variable Micropipettes
Catalog #: 470105-434
Supplier: Edvotek
Specifications
- Volume:1 - 200 µl
- Description:2 racks of 96 tips - Yellow
- Supplier No.:636
Specifications
About this item
Our newly re-designed variable micropipettes are sturdily designed with volumes ranging from 0.1 to 5000 µl.
- New and improved
- High quality, same price
- More durable and ergonomic
- Lifetime warranty
- Light gray color
They are easy to use, highly accurate and use standard micropipette tips. The volume is easily selected by twisting the top. The lightweight design and tip ejector make operation fast and easy.
Ordering information: A tool and instructions are included for self-calibration.
Customers Who Bought This Also Bought