Order Entry
ContactUsLinkComponent
OTICAL BENCH 1.8M STEEL W/ ACCESSORIES
Optical Bench, 1.8 m
Catalog #: 470039-934
Supplier:  VWR International
OTICAL BENCH 1.8M STEEL W/ ACCESSORIES
Optical Bench, 1.8 m
Catalog #: 470039-934
Supplier:  VWR International

Specifications

  • Description:
    Optical Bench, 1.8 m
  • No. of Lab Groups:
    1
  • Cat. no.:
    470039-934
  • Supplier No.:
    PH0652

Specifications

About this item

This is a complete optical bench system at an affordable price.

  • Bright Light Source
  • Lenses and Color Filters Provided
  • Steel Construction

This bench, made from a bent steel U section, is built for years of classroom use.

The set includes everything you need for advanced optical experimentation.

Ordering information: This kit includes a 1.8 m optical bench track with feet supports; 5 pieces of riders; cylindrical lens on holder mounted with slit holder; 5 pieces of lens holder; a 10, 20, and 50 cm convex lenses; a 10, 20, and 50 cm concave lens; set of four color filters with slits; a 100 x 100 mm mirror; a 100 x 100 mm screen; translucent screen; slight holder; light source; and set of diaphragms. A 12V power supply is required but not included.
Customers Who Bought This Also Bought