Order Entry
ContactUsLinkComponent
STIRRING ROD GLASS 200MM L 5MM D PK12
Glass Stirring Rods
Catalog #: 470020-788
STIRRING ROD GLASS 200MM L 5MM D PK12
Glass Stirring Rods
Catalog #: 470020-788

Specifications

  • Ø:
    5 mm
  • Length:
    200 mm
  • Cat. no.:
    470020-788
  • Supplier No.:
    GSR008

Specifications

About this item

Glass stirring rod features fire polished flint glass.

  • Rounded ends

Stirring rods are made of soft flint glass, with both ends fire-polished and rounded.

Customers Who Bought This Also Bought