Order Entry
ContactUsLinkComponent
FLINT GLASS WATCH GLASS 90MM O.D. PKG/12
Flint Glass Watch Glasses
Catalog #: 470020-744
FLINT GLASS WATCH GLASS 90MM O.D. PKG/12
Flint Glass Watch Glasses
Catalog #: 470020-744

Specifications

  • Ø:
    90 mm
  • Description:
    Flint glass watch glasses
  • Cat. no.:
    470020-744
  • Supplier No.:
    WGL090

Specifications

About this item

Useful clear watch glasses for evaporation.

  • Flint glass
  • Flame polished edges

Made from clear flint glass (1.8 to 2.0mm thick), fine ground and polished edges.

Customers Who Bought This Also Bought